missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
HNRPA3 Polyclonal specifically detects HNRPA3 in Human samples. It is validated for Western Blot.
Especificaciones
Especificaciones
| Antígeno | HNRPA3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/ml |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1.0 ug/ml |
| Formulación | PBS, 2% Sucrose |
| Alias de gen | heterogeneous nuclear ribonucleoprotein A3 |
| Especie del huésped | Rabbit |
| Inmunógeno | The immunogen is a synthetic peptide directed towards the N-terminal region of Human HNRPA3 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?