missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
HNF-4 gamma/NR2A2 Polyclonal antibody specifically detects HNF-4 gamma/NR2A2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | HNF-4 gamma/NR2A2 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry, Immunohistochemistry-Paraffin 16 - 32 μg/mL |
| Formulación | PBS |
| Alias de gen | hepatocyte nuclear factor 4, gamma, hepatocyte nuclear factor 4-gamma, HNF-4-gamma, NR2A2Nuclear receptor subfamily 2 group A member 2, NR2A3 |
| Especie del huésped | Rabbit |
| Inmunógeno | Synthetic 16 amino acid peptide from internal region of human HNF-4 gamma/NR2A2. |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?