missing translation for 'onlineSavingsMsg'
Learn More

HMGB1/HMG-1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Código de producto. 18060664 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18060664 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18060664 Proveedor Novus Biologicals N.º de proveedor NB1002322AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HMGB1/HMG-1
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Clasificación Polyclonal
Conjugado Alexa Fluor 750
Dilución Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated
Formulación 50 mM sodium borate with 0.05% sodium azide
Alias de gen Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Símbolos de los genes HMGB1
Especie del huésped Rabbit
Inmunógeno A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt No.P09429]
Cantidad 0.1 mL
Primario o secundario Primary
ID de gen (Entrez) 3146
Especies diana Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.