missing translation for 'onlineSavingsMsg'
Learn More

HMG-CoA Reductase/HMGCR Antibody (CL0260), Novus Biologicals™

Código de producto. 18247991 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18247991 100 μL 100 microlitros
18661218 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 18247991 Supplier Novus Biologicals Supplier No. NBP261617

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMG-CoA Reductase/HMGCR Monoclonal specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HMG-CoA Reductase/HMGCR
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon CL0260
Conjugado Unconjugated
Dilución Western Blot 1 ug/ml, Immunohistochemistry 1:50 - 1:200
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3
Símbolos de los genes HMGCR
Especie del huésped Mouse
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: GAKFLSDAEIIQLVNAKHIPAYKLETLMETHERGVSIRRQLLSKKLSEPSSLQYLPYRDYNYSLVMGACCENVIGYMPIPVGVAGPLCLDEKEFQVPMATTEGCLVASTNRGCRAIGLGGGASSRVLADGMTRGPVVRLPRACDSAEVKAWLETSEGFAVIKEAFDSTSRFARLQKLHTSIAGRNLYIRFQSRSGDAMGMNMISKGTEKALSKLHEYFPEMQILAVSGNYCTDKKPAAINWIEGRGKSVVCEAVIPAKVVREVLKTTT
Método de purificación Protein A purified
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 3156
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.