missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.3A Antibody, Novus Biologicals™

Product Code. p-200065348 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
100 μg
50 μg
missing translation for 'unitSize'
100 microlitros
50 microlitros
Product Code. Cantidad unitSize
18607975 100 μg 100 microlitros
18672666 50 μg 50 microlitros
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18607975

missing translation for 'mfr': Novus Biologicals NBP298769100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Histone H3.3A Polyclonal antibody specifically detects Histone H3.3A in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Histone H3.3A
Aplicaciones Western Blot, Immunoprecipitation
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunoprecipitation 1-4 μL/mg of lysate
Formulación 0.2 um filtered solution in PBS
Alias de gen H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human Histone H3.3A.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Chromatin Research, Epigenetics
Primario o secundario Primary
ID de gen (Entrez) 3020
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.