missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.1 Antibody (27F2), Novus Biologicals™

Product Code. 30025313 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μL
50 μL
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30025313 50 μL 50 microlitros
30031521 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30025313 Supplier Novus Biologicals Supplier No. NBP32622850ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Histone H3.1
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Clasificación Monoclonal
Clon 27F2
Conjugado Unconjugated
Dilución Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200
Formulación PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gen H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Especie del huésped Rabbit
Inmunógeno Synthesized peptide [UniProt P68431]
Método de purificación Affinity purified
Cantidad 50 μL
Estado normativo RUO
Disciplina de investigación Epigenetics, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 8353
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.