Descripción
Histone H2AX (p Ser139) Polyclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Especificaciones
Especificaciones
| Antígeno | Histone H2AX (p Ser139) |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Inmunógeno | This Histone H2AX [p Ser139] Antibody was developed against a synthetic phospho-peptide, which represented a portion of the C-terminus of human histone H2AX surrounding phosphorylated serine 139 (GeneID 3014). |
| Estado normativo | RUO |
| Contenido y almacenamiento | Store at 4C. Do not freeze. |
For Research Use Only