missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Histone H2A.Z Monoclonal antibody specifically detects Histone H2A.Z in Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
Specifications
Specifications
| Antígeno | Histone H2A.Z |
| Aplicaciones | Western Blot, ELISA, Immunocytochemistry, Multiplex |
| Clasificación | Monoclonal |
| Clon | RM222 |
| Conjugado | PerCP |
| Dilución | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay |
| Formulación | PBS |
| Alias de gen | H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z |
| Especie del huésped | Rabbit |
| Inmunógeno | An acetyl-peptide corresponding to Histone H2A.Z [ac Lys7] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?