Descripción
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
Especificaciones
Especificaciones
| Antígeno | HIF-1 alpha (Hydroxy Pro564) |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Especies diana | Canine, Monkey |
For Research Use Only