missing translation for 'onlineSavingsMsg'
Learn More

HIF-1 alpha Antibody, Novus Biologicals™

Código de producto. 18097803 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18097803 0.025 mL 25 microlitros
18779583 - 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18097803 Proveedor Novus Biologicals N.º de proveedor NBP102160SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 6 publications

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HIF-1 alpha
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:3500, Simple Western 1:250
Formulación PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide
N.º de referencia del gen Q16665
Alias de gen AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
Símbolos de los genes HIF1A
Especie del huésped Rabbit
Inmunógeno This HIF-1 alpha Antibody was developed against a genomic sequence made to an internal portion of human HIF-1 alpha (within amino acids 400 - 550) [Uniprot# Q16665].
Peso molecular del antígeno 93 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 3091
Especificidad de la prueba Human. Reactivity with mouse and porcine reported by a customer review. Predicted to react with monkey based on 100% sequence homology. Immunogen sequence has 96% homology to bovine, 95% to porcine, 94% to rabbit, 92% to rat, 93% to mouse isoform 2, 83% to mouse isoform 1, and 89% to chicken.
Especies diana Human, Mouse, Pig, Bovine
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.