missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heterogeneous Nuclear Ribonucleoprotein (A1-like) Antibody (S05-8G1), Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Heterogeneous Nuclear Ribonucleoprotein (A1-like) Monoclonal antibody specifically detects Heterogeneous Nuclear Ribonucleoprotein (A1-like) in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | Heterogeneous Nuclear Ribonucleoprotein (A1-like) |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | S05-8G1 |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Alias de gen | heterogeneous nuclear ribonucleoprotein A1-like 2, hnRNP A1-like 2, hnRNP core protein A1-like 2, HNRNPA1L, LOC144983, MGC102957 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide of human Heterogeneous Nuclear Ribonucleoprotein (A1-like) (Uniprot # Q5TBS2) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?