missing translation for 'onlineSavingsMsg'
Learn More

HER-2 / c-erbB-2 / neu / CD340 Mouse, Clone: ERBB2/2452, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 554.40€

Specifications

Antígeno HER-2 / c-erbB-2 / neu / CD340
Clon ERBB2/2452
Concentración 200μg/mL
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
15939957
View Documents
EnquireBio
2064-MSM12-P0
20 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A View alternative products
Save Up to 
»
15949957
View Documents
EnquireBio
2064-MSM12-P1
100 μg
554.40€
100 microgramos
View alternative products
Save Up to 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Specifications

Specifications

HER-2 / c-erbB-2 / neu / CD340
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human HER-2 protein fragment (apprx. aa 311-462)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
185kDa
ERBB2/2452
Immunohistochemistry
Unconjugated
RUO
2064
IgG2a κ
Protein A/G purified from Bioreactor concentrate.
Cell Surface
SKBR-3 Cells or Breast Cancers.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.