missing translation for 'onlineSavingsMsg'
Learn More

Helicobacter Pylori Antibody (HP/212), DyLight 550 , Novus Biologicals™

Product Code. 18577435 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
0.1 mL
missing translation for 'unitSize'
0.10 ml
Product Code. Cantidad unitSize
18577435 0.1 mL 0.10 ml
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18577435

missing translation for 'mfr': Novus Biologicals NBP311570R

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. pylori samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Helicobacter Pylori
Aplicaciones Flow Cytometry, ELISA, Immunofluorescence
Clasificación Monoclonal
Clon HP/212
Conjugado DyLight 550
Formulación 50mM Sodium Borate
Alias de gen H. pylori
Especie del huésped Mouse
Inmunógeno Total sonicate of Helicobacter pylori
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
Especies diana H. pylori
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.