missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Heat Stable FGF-10 Recombinant Protein
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
50 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
5 microgramos
50 microgramos
500 microgramos
Descripción
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
Especificaciones
Especificaciones
| Número de acceso | O15520 |
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with 20 mM Na2PO4, 750 mM NaCl and no preservative; pH 7.5 |
| ID de gen (Entrez) | 2255 |
| Nombre | Heat Stable FGF-10 |
| Método de purificación | SDS-Page |
| Cantidad | 100 μg |
| Concentración de endotoxinas | 0.25 EU/μg |
| Alias de gen | AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria |
| Nombre común | FGF10 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido