missing translation for 'onlineSavingsMsg'
Learn More

Heartland Virus Glycoprotein 2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18748881 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18748881 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18748881 Supplier Novus Biologicals Supplier No. NBP241229C

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Heartland Virus Glycoprotein 2 Polyclonal antibody specifically detects Heartland Virus Glycoprotein 2 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Heartland Virus Glycoprotein 2
Aplicaciones ELISA
Clasificación Polyclonal
Conjugado DyLight 650
Formulación 50mM Sodium Borate
Alias de gen Glycoprotein polyprotein
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 19 amino acid peptide near the center of the Heartland virus glycoprotein 2. The immunogen is located within amino acids 860 - 910 of Heartland Virus Glycoprotein 2. Amino Acid Squence: VRFDQGNHGESRIYGSPLD
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Infections (Virus Bacteria and Parasites)
Primario o secundario Primary
Especies diana Virus
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.