missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HB-EGF Antibody (KHK2866) - Humanized, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
HB-EGF Monoclonal antibody specifically detects HB-EGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
Especificaciones
| Antígeno | HB-EGF |
| Aplicaciones | Flow Cytometry, ELISA, Functional Assay |
| Clasificación | Monoclonal |
| Clon | KHK2866 |
| Conjugado | Unconjugated |
| Formulación | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| N.º de referencia del gen | Q99075 |
| Alias de gen | diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor |
| Especie del huésped | Human |
| Inmunógeno | HBEGF |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?