missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Haptoglobin Antibody (HP/3834) [Alexa Fluor™ 647], Novus Biologicals™

Código de producto. 30060470 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30060470 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30060470 Proveedor Novus Biologicals™ N.º de proveedor NBP326950AF647

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Haptoglobin
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon HP/3834
Conjugado Alexa Fluor 647
Formulación 50 mM Sodium Borate
Alias de gen binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin
Especie del huésped Mouse
Inmunógeno Recombinant fragment (around aa100-250) of human Haptoglobin protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cardiovascular Biology
Primario o secundario Primary
ID de gen (Entrez) 3240
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.