missing translation for 'onlineSavingsMsg'
Learn More

HAO2 Antibody, Novus Biologicals™

Código de producto. 18332319 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.05 mg
Tamaño de la unidad:
0.05 miligramo
Product Code. Cantidad unitSize
18332319 0.05 mg 0.05 miligramo
1 options
This item is not returnable. View return policy

Product Code. 18332319

Brand: Novus Biologicals H00051179B01P

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Polyclonal Antibody

HAO2 Polyclonal antibody specifically detects HAO2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HAO2
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500
Formulación PBS (pH 7.4)
N.º de referencia del gen NP_001005783.1
Alias de gen (S)-2-hydroxy-acid oxidase, peroxisomal, Cell growth-inhibiting gene 16 protein, EC 1.1.3.15, GIG16, growth-inhibiting protein 16, HAOX2glycolate oxidase, hydroxyacid oxidase 2, hydroxyacid oxidase 2 (long chain), Long chain alpha-hydroxy acid oxidase, Long-chain L-2-hydroxy acid oxidase
Especie del huésped Mouse
Inmunógeno HAO2 (NP_001005783.1, 1 a.a. - 351 a.a.) full-length human protein. MSLVCLTDFQAHAREQLSKSTRDFIEGGADDSITRDDNIAAFKRIRLRPRYLRDVSEVDTRTTIQGEEISAPICIAPTGFHCLVWPDGEMSTARAAQAAGICYITSTFASCSLEDIVIAAPEGLRWFQLYVHPDLQLNKQLIQRVESLGFKALVITLDTPVCGNRRHDIRNQLRRNLTLTDLQSPKKGNAIPYFQMTPISTSLCWNDLSWFQSITRLPIILKGILTKEDAELAVKHNVQGIIVSNHGGRQLDEVLASIDALTEVVAAVKGKIEVYLDGGVRTGNDVLKALALGAKCIFLGRPILWGLACKGEHGVKEVLNILTNEFHTSMALTGCRSVAEINRNLVQFSRL
Método de purificación Protein A purified
Cantidad 0.05 mg
Estado normativo RUO
Disciplina de investigación Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 51179
Especies diana Human
Contenido y almacenamiento Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.