missing translation for 'onlineSavingsMsg'
Learn More

HAGHL Antibody (5B12), Novus Biologicals™

Código de producto. 18560701 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microgramos
Product Code. Cantidad unitSize
18560701 100 μg 100 microgramos
1 options
This item is not returnable. View return policy

Product Code. 18560701

Brand: Novus Biologicals H00084264M01100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HAGHL Monoclonal antibody specifically detects HAGHL in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno HAGHL
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 5B12
Conjugado Unconjugated
Formulación In 1x PBS, pH 7.4
N.º de referencia del gen NP_115680.1
Alias de gen EC 3.1.2.-, GLO2-like/ RJD12, hydroxyacyl glutathione hydrolase-like, hydroxyacylglutathione hydrolase-like, hydroxyacylglutathione hydrolase-like protein, MGC2605
Especie del huésped Mouse
Inmunógeno HAGHL (NP_115680.1, 183 a.a. ∽ 282 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QKVEPCNDHVRAKLSWAKKRDEDDVPTVPSTLGEERLYNPFLRVAEEPVRKFTGKAVPADVLEALCKERARFEQAGEPRQPQARALLALQWGLLSAAPHD
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84264
Especies diana Human
Contenido y almacenamiento Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.