missing translation for 'onlineSavingsMsg'
Learn More

H1F0 Antibody (r1415-1), Alexa Fluor™ 405, Novus Biologicals™

Código de producto. 18783312 Tienda Bio Techne Productos
Change view
Click to view available options
:
100 μg, Alexa Fluor 405
Código de producto.
18783312 100 μg, Alexa Fluor 405
1 options
This item is not returnable. View return policy

Product Code. 18783312

Brand: Novus Biologicals NBP254303AF405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

H1F0 Monoclonal antibody specifically detects H1F0 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno H1F0
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon r1415-1
Conjugado Alexa Fluor 405
Formulación 50 mM Sodium Borate
Alias de gen H1 histone family, member 0, H1.0, H1(0), H1-0, H10, H1FVMGC5241, Histone H1', Histone H1(0), histone H1.0
Especie del huésped Mouse
Inmunógeno Recombinant full-length human H1F0 protein
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3005
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.