Learn More
GSTA3 Antibody, Novus Biologicals™
Código de producto. 18604608 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18604608 100 μg 100 microlitros
18683769 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18604608 Proveedor Novus Biologicals N.º de proveedor NBP299360100ul
Rabbit Polyclonal Antibody
GSTA3 Polyclonal antibody specifically detects GSTA3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno GSTA3
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000
Formulación 0.2 um filtered solution in PBS
Alias de gen EC 2.5.1.18, glutathione S-alkyltransferase A3, glutathione S-aralkyltransferase A3, glutathione S-aryltransferase A3, glutathione S-transferase A3, Glutathione S-transferase A3-3, glutathione S-transferase alpha 3, GST class-alpha member 3, GSTA3-3, GTA3, MGC22232, S-(hydroxyalkyl)glutathione lyase A3
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GSTA3.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 2940
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG