missing translation for 'onlineSavingsMsg'
Learn More

GRB2 Antibody (M331), Novus Biologicals™

Código de producto. 18626304 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18626304 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18626304

missing translation for 'mfr': Novus Biologicals NBP323354

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GRB2 Monoclonal antibody specifically detects GRB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GRB2
Aplicaciones Western Blot, Immunofluorescence, Immunoprecipitation
Clasificación Monoclonal
Clon M331
Conjugado Unconjugated
Dilución Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:50
Formulación PBS, 1 mg/ml BSA and 50% glycerol
Alias de gen abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2
Especie del huésped Mouse
Inmunógeno Clone (M331) was generated from a full length recombinant rat Grb2 protein. This sequence has high homology to human and mouse Grb2. [UniProt# P62994]
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Breast Cancer, Cancer, Extracellular Matrix, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 2885
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.