Learn More
MedChemExpress Granzyme B/GZMB Protein, Mouse (CHO)
Especificaciones
Especificaciones
| Peso molecular | Approximately 32 kDa |
| Cantidad | 10 μg |
| Recombinante | Recombinant |
| Contenido y almacenamiento | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Condiciones de envío | Room temperature |
| Sinónimo | rMuGranzyme B∣ CTLA-1∣ CCP1∣ Fragmentin-2∣ Gzmb |