missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Granzyme B Polyclonal specifically detects Granzyme B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
| Antígeno | Granzyme B |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Clasificación | Polyclonal |
| Conjugado | DyLight 488 |
| Dilución | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT |
| Símbolos de los genes | GZMB |
| Especie del huésped | Rabbit |
| Inmunógeno | Partial recombinant protein made to the human Granzyme B protein (amino acids 19-247) [UniProt P10144] |
| Mostrar más |
For Research Use Only (RUO)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?