missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR55 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55249-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. .Especificaciones
| GPR55 | |
| Polyclonal | |
| Unconjugated | |
| GPR55 | |
| G protein coupled receptor 55, GPR55 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9290 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2T6 | |
| GPR55 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido