Learn More
Goat anti-Canine IgM Heavy Chain Secondary Antibody, Biotin, Novus Biologicals™
Código de producto. 30228465 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mL
Tamaño de la unidad:
1 ml
30228465 1 mL 1 ml
1 options
Código de producto. 30228465
Marca: Novus Biologicals NB7318B
Goat Polyclonal Antibody
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IgM Heavy Chain
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Especie del huésped Goat
Inmunógeno Dog IgM
Método de purificación Immunogen affinity purified
Cantidad 1 mL
Estado normativo RUO
Disciplina de investigación B Cell Development and Differentiation Markers, Immunology
Primario o secundario Secondary
ID de gen (Entrez) 3507
Especies diana Canine
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG