Learn More
Abnova™ GMEB2 Recombinant Protein
Marca:  Abnova™ H00026205-P01.25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16168162
  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock
Descripción
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
Especificaciones
AAH36305
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
84.04
8
Glutathione Sepharose 4 Fast Flow
25 ug
MATPDVSVHMEEVVVVTTPDTAVDGSGVEGVKTVLVTTNLAPHGGDLTEDNMETENAAAAAAAAFTASSQLKEAVLVKMAEEGENLEAEIVYPITCGDSRANLIWRKFVCPGINVKCVQYDEHVISPKEFVHLAGKSTLKDWKRAIRMNGIMLRKIMDSGELDFYQHDKVCSNTCRSTKIDLSGARVSLSSPTSAEYIPLTPAAADVNGSPATITIETCEDPGDWTAAIGDDTFTFWRGLKDAGLLDEVIQEFHQELVETMRGLQQRVQDPPLQLRDAVLLNNIVQNFGMLDLVKKVLASHKCQMDRSREQYARDLAALEQQCDEHRRRAKELKHKSQHLSNVLMTLTPVSLPPPVKRPRLARATSGPAAMASQVLTQSAQLALGPGVPVPQLTSVPLGKVVSTLPSTVLGKGSLQAPPASSPASPLLGGYTVLASSGSTYPSTVEIHPDASSLTVLSTAAVQDGSTVFKVVSPLQLLTLPGLGPTLQNVAQASPGSSTIVTVPAGAAPGPEEHTATIEVAAMAEDHERK
KIAA1269/P79PIF/PIF79
GMEB2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
26205
GMEB2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GMEB2
Human
Recombinant
Solution