missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2 Ganglioside Human anti-Human, Clone: L55-81, Novus Biologicals™
Human Monoclonal Antibody
Marca: Bio-Techne NBP2-81277-0.05MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Cell depletion
Especificaciones
GM2 Ganglioside | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Depletion Assay | |
GM2 (ganglioside), Tay-Sachs ganglioside | |
L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. | |
0.05 mg | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Depletion Assay | |
L55-81 | |
Unconjugated | |
PBS | |
Human | |
Affinity purified | |
RUO | |
Human | |
IgM κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
GM2 Ganglioside Human anti-Human, Clone: L55-81, Novus Biologicals™ > 0.05 mg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido