missing translation for 'onlineSavingsMsg'
Learn More

GM-CSF Antibody (6804), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 30059569 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30059569 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30059569

Brand: Novus Biologicals FAB615G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno GM-CSF
Aplicaciones ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 6804
Conjugado Alexa Fluor 488
Dilución ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulación 50mM Sodium Borate
Alias de gen colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human GM-CSF, Ala18-Glu144, Accession # P04141
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Cytokine Research, Immunology
Primario o secundario Primary
ID de gen (Entrez) 1437
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.