missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Glypican 5 Polyclonal antibody specifically detects Glypican 5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Glypican 5 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulación | PBS (pH 7.2), 40% Glycerol |
| Alias de gen | bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: AELNPHWHAYIRSLEELSDAMHGTYDIGHVLLNFHLLVNDAVLQAHLNGQKLLEQVNRICGRPVRTPTQSPRCSFDQSKEKHGMKTTTRN |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?