missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A Antibody (YTH89.1), Janelia Fluor™ 646, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
| Antígeno | GlycophorinA |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | YTH89.1 |
| Conjugado | Janelia Fluor 646 |
| Dilución | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR |
| Símbolos de los genes | GYPA |
| Especie del huésped | Rat |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?