missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Glutathione S-transferase Mu 5 |
| Aplicaciones | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 μL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 |
| Formulación | 0.2 um filtered solution in PBS |
| Alias de gen | EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with purified, recombinant Human Glutathione S-transferase Mu 5 (Accession#: AAH58881.1; Met1-Lys218) |
| Método de purificación | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?