Learn More
GLUT5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27571-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SLC2A5 (GLUT5) is a cytochalasin B-sensitive carrier. It seems to function primarily as a fructose transporter.Especificaciones
| GLUT5 | |
| Polyclonal | |
| Unconjugated | |
| SLC2A5 | |
| Fructose transporter, GLUT 5, GLUT5, SLC2A5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56485, 6518 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P22732, Q9WV38 | |
| SLC2A5 | |
| GLUT5 Fusion Protein Ag25682 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |