missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532422
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control is stomach tissue.
This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.Especificaciones
Gastrin | |
Polyclonal | |
PBS with 1% BSA and <0.1% sodium azide; pH 7.20 | |
P01350 | |
Gast | |
Synthetic peptide derived from N-terminus of gastrin 1 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Gast | |
Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |