Learn More
GAPDH Antibody, Novus Biologicals™
Código de producto. 18033316 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
18033316 0.025 mL 25 microlitros
18709753 0.1 mL 0.10 ml
2 options
Código de producto. 18033316
Marca: Novus Biologicals NB3003270.025ML
Rabbit Polyclonal Antibody has been used in 33 publications
GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Chicken, Equine, Fungi, Invertebrate, Yeast samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno GAPDH
Aplicaciones Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:30000, Simple Western 1:500, Immunohistochemistry 1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Formulación Whole antisera with 0.05% Sodium Azide
N.º de referencia del gen P10096
Alias de gen aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Símbolos de los genes GAPDH
Especie del huésped Rabbit
Inmunógeno This GAPDH antibody was developed against full length recombinant human GAPDH
Peso molecular del antígeno 36 kDa
Método de purificación Unpurified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primario o secundario Primary
ID de gen (Entrez) 2597
Especificidad de la prueba Human, cow, pig, mouse, rat, bird and chicken. Yeast reactivity reported in scientific literature (PMID: 23709183). Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Invertebrate reactivity reported in scientific literature (PMID: 18638509).
Especies diana Rat, Chicken, Mouse, Pig, Yeast, Equine, Fungi, Bovine, Invertebrate, Bacteria, Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Antisera
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only