Learn More
GAPDH Antibody (1A10) - BSA Free, Novus Biologicals™
Código de producto. 18064105 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
18064105 0.025 mL 25 microlitros
18759953 - 0.10 ml
2 options
Código de producto. 18064105
Marca: Novus Biologicals NBP147339SS
Mouse Monoclonal Antibody has been used in 5 publications
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno GAPDH
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 1A10
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 5 ug/ml, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
N.º de referencia del gen P04406
Alias de gen aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Símbolos de los genes GAPDH
Especie del huésped Mouse
Inmunógeno GAPDH antibody developed against purified recombinant fragment of human GAPDH expressed in E. coli. [UniProt# P04406]
Peso molecular del antígeno 36 kDa
Método de purificación Ammonium sulfate precipitation
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primario o secundario Primary
ID de gen (Entrez) 2597
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Precipitation
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only