Learn More
gamma catenin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 27872-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16815885
  • 410.00€ / 150 microlitros
Descripción
JUP is a major cytoplasmic protein which is the only known constituent common to submembranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
Especificaciones
Especificaciones
gamma catenin
Polyclonal
Unconjugated
JUP
ARVD12, Catenin gamma, CTNNG, Desmoplakin 3, Desmoplakin III, DP3, DPIII, junction plakoglobin, JUP, PDGB, PKGB, gamma-Catenin
Rabbit
Antigen Affinity Chromatography
RUO
3728
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14923
JUP
gamma catenin Fusion Protein Ag27394
150 μL
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones