Learn More
Galectin 10 Mouse anti-Human, Unconjugated, Clone: 5D9B7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67581-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Galectin 10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Galectin 10. This antibody recognizes human antigen. The Galectin 10 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| Galectin 10 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 5D9B7 | |
| Unconjugated | |
| 150 μL | |
| 1178 | |
| IgG1 |