Learn More
Novus Biologicals™ GAD2/GAD65 Antibody (U.Washington patent anti-GAD65), Novus Biologicals™
Código de producto. 30046376 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
30046376 50 μg 50 microgramos
30047531 1 mg 1 miligramo
30047105 100 μg 100 microgramos
3 options
Código de producto. 30046376
Marca: Novus Biologicals™ NBP32868850ug
This item is not returnable. View return policy
Human Monoclonal Antibody
GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Antígeno GAD2/GAD65
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon U.Washington patent anti-GAD65
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q05329
Alias de gen EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase
Especie del huésped Human
Inmunógeno GAD65
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 2572
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1