missing translation for 'onlineSavingsMsg'
Learn More

GAD1 / GAD67 (GABAergic Neuronal Marker) Mouse, Clone: GAD1/2391, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 554.40€

Especificaciones

Antígeno GAD1 / GAD67 (GABAergic Neuronal Marker)
Clon GAD1/2391
Concentración 200μg/mL
Aplicaciones ELISA, Western Blot, Immunohistochemistry
Clasificación Monoclonal
Ver más especificaciones

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
15910078
View Documents
EnquireBio
2571-MSM1-P0
20 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A View alternative products
Save Up to 
»
15920078
View Documents
EnquireBio
2571-MSM1-P1
100 μg
554.40€
100 microgramos
View alternative products
Save Up to 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Specifications

Specifications

GAD1 / GAD67 (GABAergic Neuronal Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human GAD1 (GAD67) protein fragment (apprx. aa 72-135)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
67kDa
GAD1/2391
ELISA, Western Blot, Immunohistochemistry
Unconjugated
RUO
2571
IgG1 κ
Protein A/G purified from Bioreactor concentrate.
Cytoplasmic
K-562 or HEK293 Cells. Pancreas
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.