Learn More
Novus Biologicals™ FZR1/CDH1 Antibody (Stem Centrx patent anti-Cadherin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28089-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FZR1/CDH1 Monoclonal antibody specifically detects FZR1/CDH1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
| FZR1/CDH1 | |
| Monoclonal | |
| Unconjugated | |
| P12830 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Stem Centrx patent anti-Cadherin-1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog | |
| CDH1/ E-cadherin/ CD324 | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction | |
| 51343 | |
| Human | |
| Purified |