missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
218.00€ - 520.00€
Especificaciones
| Antígeno | FTO |
|---|---|
| Concentración | 1.1 mg/mL |
| Dilución | Western Blot 0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 28205605), Immunoblotting reported in scientific literature (PMID 22019092) |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunoblot |
| Clasificación | Polyclonal |
Descripción
FTO Polyclonal specifically detects FTO in Human, Mouse, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoblotting.Especificaciones
| FTO | |
| Western Blot 0.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 28205605), Immunoblotting reported in scientific literature (PMID 22019092) | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| FTO | |
| IgG | |
| Affinity Purified | |
| 60 kDa |
| 1.1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoblot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Chicken, Primate | |
| Q9C0B1 | |
| 79068 | |
| A synthetic peptide within the C-terminal region (residues 400-505) of the human FTO protein. [Swiss-Prot# Q9C0B1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto