missing translation for 'onlineSavingsMsg'
Learn More

Frataxin Mouse anti-Human, Clone: 1.00E+12, Novus Biologicals™

Product Code. 18212562 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
50 μL
100 μL
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18212562 100 μL 100 microlitros
18203502 50 μL 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18212562 Brand Novus Biologicals Brand No. NBP259478100UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Frataxin
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 1000000000000
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:1000, Flow Cytometry 1:10-1:1076, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulación Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Alias de gen CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Símbolos de los genes FXN
Especie del huésped Mouse
Inmunógeno Recombinant human Frataxin (42-210aa) purified from E. coli
Método de purificación Protein A purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Diabetes Research
Primario o secundario Primary
ID de gen (Entrez) 2395
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.