Learn More
Eppendorf Flask Clamp for Incubator Shaker CS220, HTD Compatible
Código de producto. 30280813 Tienda Eppendorf Productos
Click to view available options
Admite:
Matraces de 100/125 mL
Matraces de 1 L
Matraces de 2,8 L
Matraces de 200/250 mL
Matraces de 2 L
Matraces de 3/5 L
Matraces de 500 mL
Tamaño de la unidad:
cada uno
Código de producto. 30280813
Marca: Eppendorf 6740070160
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Eppendorf flask clamps secure a wide range of vessels to the shaker platform and are constructed of a single piece stainless-steel for added strength and safety. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
  • For Erlenmeyer flasks (standard, narrow/wide neck), Corning™, Thomson™, or Nalgene™
  • Fernbach
  • 180°C resistant
  • Autoclavable
  • Stainless steel
  • Includes M4x7 screws
TRUSTED_SUSTAINABILITY
Especificaciones
Admite Matraces de 200/250 mL
Tipo de producto Lab Shaker Clamp
Material Stainless Steel
Para utilizar con (equipo) CellXpert CS220 CO2 Incubator Shaker
Incluye M4x7 screws
Cantidad 1 Ea.