missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20425-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Fibulin-1 is a modular glycoprotein component of the elastic extracellular matrix fibers, basement membranes and blood. Fibulin-1 self associates as well as binds to calcium, fibronectin, laminin, nidogen and fibrinogen. These interactions, individually or in combination, may account for the observed association of Fibulin-1 with basement membranes, connective tissue, elastic fibers and fibrin clots. Fibulin-1 expression is stimulated by estrogen in ovarian cancer cell lines and has been suggested as both an agent of metastasis in ovarian cancer cells and an indicator for predicting cancer risk or aggressiveness in ovarian carcinomas. Other studies point to the inhibition of cancer cell motility with increasing exposure to Fibulin-1. The exact function of Fibulin-1 in the cell is unknown.Especificaciones
| Fibulin-1 | |
| Polyclonal | |
| Unconjugated | |
| FBLN1 | |
| FBLN, FBLN1, FIBL 1, Fibulin-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14114, 2192, 315191 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23142, Q08879 | |
| Fbln1 | |
| Fibulin-1 Fusion Protein Ag14249 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido