missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | FGF18 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL7628 |
| Conjugado | Janelia Fluor 549 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FGF-18ZFGF5, fibroblast growth factor 18, zFGF5 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a synthetic peptide derived from O76093, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?