missing translation for 'onlineSavingsMsg'
Learn More

FGF-13 Antibody (298923), DyLight 680, Novus Biologicals™

Código de producto. 18583068 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18583068 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18583068

Brand: Novus Biologicals FAB1909Y

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno FGF-13
Aplicaciones Western Blot, Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 298923
Conjugado DyLight 680
Dilución Western Blot, Flow Cytometry, CyTOF-ready
Formulación 50mM Sodium Borate
Alias de gen FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human FGF-13 isoform 1B, Ala2-Thr192, Accession # NP_378668
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Biologically Active Proteins, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 2258
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.