Descripción
Ferroportin/SLC40A1 Polyclonal specifically detects Ferroportin/SLC40A1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Especificaciones
Especificaciones
| Antígeno | Ferroportin/SLC40A1 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Especie del huésped | Rabbit |
| Cantidad | 0.1 mL |
| Especies diana | Human, Mouse, Pig, Bovine |
For Research Use Only