Learn More
Fc gamma RIIA/CD32a Antibody (S09-5D1), Novus Biologicals™
Código de producto. 18696694 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18696694 50 μg 50 microlitros
18661834 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18696694 Proveedor Novus Biologicals N.º de proveedor NBP31994750ul
Rabbit Monoclonal Antibody
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Fc gamma RIIA/CD32a
Aplicaciones Western Blot
Clasificación Monoclonal
Clon S09-5D1
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human Fc gamma RIIA/CD32a (Uniprot # P12318)
Método de purificación Affinity purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 2212
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG