Learn More
Fc gamma RIIA/CD32a Antibody (IV.3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Código de producto. 18650294 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.2 mg
Tamaño de la unidad:
0.02 miligramo
0.20 miligramo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18650294 0.025 mg 0.02 miligramo
18657134 0.2 mg 0.20 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18650294 Proveedor Novus Biologicals N.º de proveedor NBP3201590.025mg
Rabbit Monoclonal Antibody
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Inhibition Activity, Neutralization
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Fc gamma RIIA/CD32a
Aplicaciones Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays
Clasificación Monoclonal
Clon IV.3
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS
Alias de gen CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Especie del huésped Rabbit
Inmunógeno The original antibody was generated against the FcR of K562 cells.
Método de purificación Protein A purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 2212
Especies diana Human
Contenido y almacenamiento Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Formulario Purified
Isotype IgG κ